
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate) ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:64987-85-5
:
Formula:C₁₆H₁₈N₂O₆
MDL Number:MFCD00009634
:334.33 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):8.20 to 8.50 %
- Melting point:175.0 to 179.0 deg-C
Specification Test Results
Frequently Bought Together