Sodium [(R)-1,2-diaminopropane-N,N,N',N'-tetraacetato]samarate(III) ≥90.0% (by titrimetric analysis)
TCIAS0473-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (Chelometric Titration):min. 90.0 %
- Specific rotation [a]20/D:-4.0 to -6.0 deg (C=1, pH 10.0 Buffer sol.)
- Functionality test:to pass test
:
:475.59 g/mol
:C₁₁H₁₅N₂O₈Sm
:MFCD06797164