

Salcomine [=N,N'-bis(salicylidene)ethylenediiminocobalt(II)] ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:14167-18-1
:
Formula:C₁₆H₁₆CoN₂O₂
MDL Number:MFCD00000009
:327.24 g/mol
EINECS:238-012-5
:GHS07
:GHS08
Specification Test Results
- Appearance:Orange to Amber to Dark red powder to crystal
- Purity (Chelometric Titration):min. 95.0 %
Specification Test Results
Related Information
Frequently Bought Together