Order Entry
Italy
ContactUsLinkComponent
Methyl sorbate ≥98.0% (by GC)
Methyl sorbate ≥98.0% (by GC)
  TCIAS0056-25ML
 :  TCI
 :  
Methyl sorbate ≥98.0% (by GC)
  TCIAS0056-25ML
 :  TCI
 :  

 

  • Pk:
    25 mL
  • Seq:
    2315
  • Environmentally Preferable:

 

 

  • Appearance:
    White or Colorless to Light yellow powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 : Sorbic Acid Methyl Ester
 :
 :126.16 g/mol
 :216-160-1
 :1515-80-6
 :C₇H₁₀O₂
 :MFCD00038319
 :174°C