Order Entry
Italy
ContactUsLinkComponent
Di-n-octyl sebacate ≥95.0% (by GC)
Di-n-octyl sebacate ≥95.0% (by GC)
Catalog # TCIAS0028-25ML
Supplier:  TCI
CAS Number:  
Di-n-octyl sebacate ≥95.0% (by GC)
Catalog # TCIAS0028-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    White or Colorless to Light yellow powder to lump to clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :426.68 g/mol
Boiling Pt:225°C
EINECS:238-499-4
CAS Number:14491-66-8
Formula:C₂₆H₅₀O₄
MDL Number:MFCD00059269