Rasagiline mesylate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:161735-79-1
:
Formula:C₁₂H₁₃N·CH₄O₃S
MDL Number:MFCD08460604
:267.34 g/mol
Merck Index:14,08116
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 95.0 %
- Melting point:155.0 to 159.0 deg-C
- Specific rotation [a]20/D:+20.0 to +22.0 deg (C=2, ethanol)
Specification Test Results
Frequently Bought Together