Rifabutin ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:72559-06-9
:
Formula:C₄₆H₆₂N₄O₁₁
MDL Number:MFCD00866816
:847.02 g/mol
Merck Index:14,08213
Storage Temperature:Freezer
Specification Test Results
- Appearance:Red to Dark red to Brown powder to crystaline
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together