Ruthenium 5% on carbon wetted with ca. 40% water
Supplier: TCI
Certificates
About this item
CAS Number:7440-18-8
:
Formula:Ru
MDL Number:MFCD00011207
:101.07 g/mol
Merck Index:14,08299
EINECS:231-127-1
Specification Test Results
- Appearance:Black powder to crystal
- Content (Ruthenium):4.9 to 5.1 % (on dry base)
Specification Test Results
Related Information
Frequently Bought Together