Order Entry
Italy
ContactUsLinkComponent
 
Reserpine ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
Reserpine ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    -172 to -182 deg (C=1, DMF)

 

Frequently Bought Together