Order Entry
Italy
ContactUsLinkComponent
 
 
2-Phenylpyridine 1-oxide ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
 
2-Phenylpyridine 1-oxide ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    156.0 to 160.0 deg-C

 

Frequently Bought Together