N-Phenyl-2-hexyl[60]fulleropyrrolidine ≥98.0% (by HPLC)
Catalog # TCIAP2744-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Orange to Amber to Dark red powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
About this item
:
:923.99 g/mol
CAS Number:1426332-00-4
Formula:C₇₄H₂₁N