1-[1-Phenyl-3,4-dihydro-1H-pyrido[3,4-b]indol-2(9H)-yl]pentan-1-one ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:334939-35-4
:
Formula:C₂₂H₂₄N₂O
MDL Number:MFCD01496905
:332.45 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):8.00 to 8.90 %
- Melting point:187.0 to 191.0 deg-C
Specification Test Results
Frequently Bought Together