Order Entry
Italy
ContactUsLinkComponent
 
4,7-Phenanthroline ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
4,7-Phenanthroline ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    172.0 to 176.0 deg-C

 

Frequently Bought Together