5-Phenyl-5H-pyrido[3,2-b]indole ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1541200-53-6
:
Formula:C₁₇H₁₂N₂
:244.30 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:99.0 to 103.0 deg-C
Specification Test Results
Frequently Bought Together