Order Entry
Italy
ContactUsLinkComponent
PD 169316 ≥98.0% (by HPLC)
PD 169316 ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
PD 169316 ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Elemental analysis (Nitrogen):
    14.70 to 16.30 %

Specification Test Results

Frequently Bought Together