2-Oleoyl-1-palmitoyl-Sn-glycero-3-phospho-rac-(1-glycerol)sodium salt ≥98.0% (by titrimetric analysis)
TCIAP2522-250MG
: TCI
:
- Pk:250 mg
- Seq:2590
- Appearance:White to Almost white powder to crystal
- Purity (HPLC) (CAD):min. 94.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
:
:771.002 g/mol
:Freezer
:268550-95-4
:C₄₀H₇₆NaO₁₀P
:MFCD00674290