
5-Phenyl-5,12-dihydroindolo[3,2-a]carbazole ≥98.0% (by HPLC)
Catalog # TCIAP2498-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Melting point:168.0 to 172.0 deg-C
Specification Test Results
About this item
:
:332.41 g/mol
:GHS07
CAS Number:1247053-55-9
Formula:C₂₄H₁₆N₂
MDL Number:MFCD28975052