5H-Pyrido[4,3-b]indole ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:244-69-9
:
Formula:C₁₁H₈N₂
MDL Number:MFCD00965976
:168.20 g/mol
Merck Index:14,01805
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:231.0 to 235.0 deg-C
Specification Test Results
Frequently Bought Together