
1,4-Dioxa-8-azaspiro[4.5]decane ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:177-11-7
:
Formula:C₇H₁₃NO₂
MDL Number:MFCD00005976
:143.186 g/mol
Boiling Pt:110°C
Flash Pt:81°C
EINECS:205-868-6
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together