
1,3-Propylenediphosphonic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4671-82-3
:
Formula:C₃H₁₀O₆P₂
MDL Number:MFCD00050066
:204.06 g/mol
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:170.0 to 177.0 deg-C
Specification Test Results
Frequently Bought Together