Order Entry
Italy
ContactUsLinkComponent
1,5-Pentylenediphosphonic acid ≥98.0% (by titrimetric analysis)
Corrosive
1,5-Pentylenediphosphonic acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAP2300-1G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
1,5-Pentylenediphosphonic acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAP2300-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    154.0 to 158.0 deg-C

Specification Test Results

About this item

 :
 :232.11 g/mol
UN:3261
ADR:8,III
 :GHS05
CAS Number:4672-25-7
Formula:C₅H₁₄O₆P₂
MDL Number:MFCD00800268