
4-(2-Phenyl-1H-benzimidazol-1-yl)phenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAP2282-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):98.0 to 106.0 %
Specification Test Results
About this item
:
:314.15 g/mol
:GHS07
CAS Number:867044-33-5
Formula:C₁₉H₁₅BN₂O₂
MDL Number:MFCD11977298