Order Entry
Italy
ContactUsLinkComponent
1,2-Diphenyl-1-butanone ≥98.0% (by GC)
1,2-Diphenyl-1-butanone ≥98.0% (by GC)
Catalog # TCIAP2269-5G
Supplier:  TCI
CAS Number:  
undefined
1,2-Diphenyl-1-butanone ≥98.0% (by GC)
Catalog # TCIAP2269-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    54.0 to 58.0 deg-C

Specification Test Results

About this item

 :
 :224.3000 g/mol
Boiling Pt:324°C
CAS Number:16282-16-9
Formula:C₁₆H₁₆O
MDL Number:MFCD00010344