
Pentafluorophenyl trifluoromethanesulphonate ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:60129-85-3
:
Formula:C₇F₈O₃S
MDL Number:MFCD07784319
:316.12 g/mol
Boiling Pt:40°C
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together