Order Entry
Italy
ContactUsLinkComponent
Propyne ∼4% ∼1.0 mol/L in DMF
Flammable
Corrosive
Toxic
Health
Propyne ∼4% ∼1.0 mol/L in DMF
Supplier:  TCI
Certificates
undefined
Flammable
Corrosive
Toxic
Health
Propyne ∼4% ∼1.0 mol/L in DMF
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (HNMR):
    4.1 to 5.3 w/w%

Specification Test Results

Frequently Bought Together