Order Entry
Italy
ContactUsLinkComponent
(R)-1-Phenyl-1,2-ethanediyl bis[4-(trans-4-pentylcyclohexyl)benzoate] ≥98.0% (by HPLC)
(R)-1-Phenyl-1,2-ethanediyl bis[4-(trans-4-pentylcyclohexyl)benzoate] ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
undefined
(R)-1-Phenyl-1,2-ethanediyl bis[4-(trans-4-pentylcyclohexyl)benzoate] ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Elemental analysis (Carbon):
    79.00 to 84.00 %
  • Melting point:
    130.0 to 134.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together