Order Entry
Italy
ContactUsLinkComponent
4-[(1-Pyrrolidinylsulphonyl)methyl]aniline ≥97.0% (by HPLC, titration analysis)
Irritante
4-[(1-Pyrrolidinylsulphonyl)methyl]aniline ≥97.0% (by HPLC, titration analysis)
Nome del fornitore:  TCI
Certificati
undefined
Irritante
4-[(1-Pyrrolidinylsulphonyl)methyl]aniline ≥97.0% (by HPLC, titration analysis)
Nome del fornitore:  TCI

Risultati dei test sulle specifiche

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Melting point:
    176.0 to 180.0 deg-C

Risultati dei test sulle specifiche

Frequently Bought Together