Order Entry
Italy
ContactUsLinkComponent
p-Xylene-bis(N-pyridinium bromide) (DPX) ≥95.0% (by HPLC, titration analysis)
p-Xylene-bis(N-pyridinium bromide) (DPX) ≥95.0% (by HPLC, titration analysis)
Catalog # TCIAP2141-250MG
Supplier:  TCI
CAS Number:  
undefined
p-Xylene-bis(N-pyridinium bromide) (DPX) ≥95.0% (by HPLC, titration analysis)
Catalog # TCIAP2141-250MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystaline
  • Purity (HPLC):
    min. 95.0 area%
  • Purity (Nonaqueous Titration):
    min. 95.0 %

Specification Test Results

About this item

Synonyms: 1,1'-(p-Xylylene)bis(1-pyridinium) dibromide, 1,1'-(1,4-Phenylenebis(methylene))bis(pyridin-1-ium) bromide, DPX, 1,1'-(p-Xylylene)bis(1-pyridinium bromide), 1,1'-[1,4-Phenylenebis(methylene)]bis(1-pyridinium) dibromide
 :
 :422.16 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:14208-10-7
Formula:C₁₈H₁₈Br₂N₂
MDL Number:MFCD00043749