Order Entry
Italy
ContactUsLinkComponent
(S)-4-Phenyl-3-[5-(4-fluorophenyl)-5-oxopentanoyl]-2-oxazolidinone ≥98.0% (by HPLC, total nitrogen)
(S)-4-Phenyl-3-[5-(4-fluorophenyl)-5-oxopentanoyl]-2-oxazolidinone ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI
Certificates
undefined
(S)-4-Phenyl-3-[5-(4-fluorophenyl)-5-oxopentanoyl]-2-oxazolidinone ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    90.0 to 94.0 deg-C
  • Specific rotation [a]20/D:
    +49.0 to +54.0 deg (C=0.2, CHCL3)

Specification Test Results

Frequently Bought Together