
Palladium(II) acetate trimer ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:53189-26-7
:
Formula:C₁₂H₁₈O₁₂Pd₃
:673.52 g/mol
EINECS:258-424-9
:GHS07
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together