
Penfluridol ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:26864-56-2
:
Formula:C₂₈H₂₇ClF₅NO
MDL Number:MFCD00866714
:523.97 g/mol
Merck Index:14,07087
Storage Temperature:Refrigerator
EINECS:248-074-5
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:106.0 to 110.0 deg-C
Specification Test Results
Frequently Bought Together