Order Entry
Italy
ContactUsLinkComponent
Phenylmagnesium bromide 16% ∼1 mol/L in THF
Flammable
Corrosive
Irritant
Health
Phenylmagnesium bromide 16% ∼1 mol/L in THF
Catalog # TCIAP2025-250G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Health
Phenylmagnesium bromide 16% ∼1 mol/L in THF
Catalog # TCIAP2025-250G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Concentration (sec Butanol method):
    16.0 to 18.5 w/w%

Specification Test Results

About this item

 :
 :181.31 g/mol
EINECS:202-867-2
UN:2924
ADR:3,II
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:100-58-3
Formula:C₆H₅BrMg
MDL Number:MFCD00000038