
10-Phenyl-9-anthraceneboronic acid (contains varying amounts of Anhydride)
TCIAP1984-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
:
:298.15 g/mol
:GHS07
:334658-75-2
:C₂₀H₁₅BO₂
:MFCD11111989