Palladium on poly(methylphenyl)silane / alumina
Catalog # TCIAP1944-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light red to Green powder to crystal
- Palladium:min.0.37% (35micro_mol/g)
Specification Test Results
About this item
:
:106.420 g/mol
:GHS07
:GHS08
CAS Number:7440-05-3
Formula:Pd
MDL Number:MFCD00011167