
Propyl methanesulphonate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1912-31-8
:
Formula:C₄H₁₀O₃S
MDL Number:MFCD00007561
:138.18 g/mol
Boiling Pt:110°C
EINECS:217-619-9
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together