3-Phenylimidazo[1,5-a]pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:35854-46-7
:
Formula:C₁₃H₁₀N₂
:194.24 g/mol
Boiling Pt:140°C
Specification Test Results
- Appearance:Light yellow to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:108.0 to 112.0 deg-C
Specification Test Results
Frequently Bought Together