
2-(1-Piperazinylcarbonyl)-1,4-benzodioxane ≥95.0% (by GC, titration analysis)
Catalog # TCIAP1836-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Nonaqueous Titration):min. 95.0 %
- Melting point:103.0 to 107.0 deg-C
Specification Test Results
About this item
:
:248.28 g/mol
:GHS07
CAS Number:70918-00-2
Formula:C₁₃H₁₆N₂O₃
MDL Number:MFCD01837119