Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Palladium
Palladium 0.1 mmol/g on SH silica gel
Palladium 0.1 mmol/g on SH silica gel
TCIAP1786-1G
:
TCI
:
Palladium 0.1 mmol/g on SH silica gel
TCIAP1786-1G
:
TCI
:
Pk:
1 g
Seq:
3047
Appearance:
White to Yellow to Orange powder to crystal
Loading:
0.1 to 0.2 mmol/g
:
:
106.420 g/mol
:
231-115-6
:
7440-05-3
:
Pd
:
MFCD00011167