Order Entry
Italy
ContactUsLinkComponent
Polyethylene glycol monododecyl ether n=∼25 for biochemical research
Irritant
Environment
Polyethylene glycol monododecyl ether n=∼25 for biochemical research
Supplier:  TCI
Certificates
undefined
Irritant
Environment
Polyethylene glycol monododecyl ether n=∼25 for biochemical research
Supplier:  TCI
Specification Test Results
  • Appearance:
    White powder to lump
  • Freezing point:
    35.0 to 40.0 deg-C
  • Acid Value:
    max. 1.0
  • The average value of n:
    23.0 to 27.0
  • Suitability for protein-analysis:
    to pass test