Order Entry
Italy
ContactUsLinkComponent
 
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer ≥95.0% (by titrimetric analysis)
  TCIAP1763-1G
 :  TCI
 :  
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer ≥95.0% (by titrimetric analysis)
  TCIAP1763-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (Argentmetric Titration):
    min. 95.0 %

 

 

 :
 :796.69 g/mol
 :12354-84-6
 :C₂₀H₃₀Cl₄Ir₂
 :MFCD00075435