
Pyridine hydrobromide ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAP1666-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:216.0 to 221.0 deg-C
Specification Test Results
About this item
:
:160.01 g/mol
Melting Pt:217°C
EINECS:242-600-7
:GHS07
CAS Number:18820-82-1
Formula:C₅H₅N
MDL Number:MFCD00012801