Order Entry
Italy
ContactUsLinkComponent
4-(4-Pyridyl)morpholine ≥98.0% (by GC, titration analysis)
Irritante
4-(4-Pyridyl)morpholine ≥98.0% (by GC, titration analysis)
Nome del fornitore:  TCI
Certificati
undefined
Irritante
4-(4-Pyridyl)morpholine ≥98.0% (by GC, titration analysis)
Nome del fornitore:  TCI

Risultati dei test sulle specifiche

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    105.0 to 109.0 deg-C

Risultati dei test sulle specifiche

Frequently Bought Together