Palladium 10% on carbon wetted with ca. 55% water
Supplier: TCI
Certificates
About this item
CAS Number:7440-05-3
:
Formula:Pd
MDL Number:MFCD00011167
:106.420 g/mol
Merck Index:14,06989
EINECS:231-115-6
Specification Test Results
- Appearance:Black powder to crystal
- Palladium:9.8 to 10.2 % (calcd.on dried substance)
Specification Test Results
Related Information
Frequently Bought Together