Poly[N-isopropylacrylamide-co-4-(diphenylphosphino)styrene]palladium(II) dichloride(ratio, acrylamide:phosphine=20:2)
Catalog # TCIAP1425-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Black powder to crystal
- Functionality test:to pass test
Specification Test Results
About this item
:
MDL Number:MFCD04041046