Poly(styrene) on resin, cross-linked with 1% DVB (200-400 mesh)
Codice catalogo TCIAP1379-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white granule
- Grain size:200 to 400 mesh
Specification Test Results
About this item
:
:125000 - 250000 g/mol
CAS Number:9003-53-6
Formula:C₈H₈
MDL Number:MFCD00084450