Pentamethylenebis[4-(10,15,20-triphenylporphyrin-5-yl)benzoate]dizinc(II) ≥97.0% (by HPLC) reagent for application of the exciton chirality method
: TCI
:210769-64-5
:
:C₉₅H₆₄N₈O₄Zn₂
:MFCD02093489
:1512.37 g/mol
:Refrigerator
- Appearance:Dark purple powder to crystal
- Purity (HPLC):min. 97.0 area%
Frequently Bought Together