Order Entry
Italy
ContactUsLinkComponent
 
 
 
 
 
Pentadecylmagnesium bromide ∼15% ∼0.4 mol/L in THF
  TCIAP1251-250G
 :  TCI
 :  
 
 
 
 
Pentadecylmagnesium bromide ∼15% ∼0.4 mol/L in THF
  TCIAP1251-250G
 :  TCI
 :  

 

  • Pk:
    250 g
  • Seq:
    3545

 

 

  • Concentration (sec Butanol method):
    15.0 to 17.0%

 

 

 :
 :315.62 g/mol
 :2924
 :3,II
 :GHS02
 :GHS05
 :GHS07
 :GHS08
 :78887-70-4
 :C₁₅H₃₁BrMg
 :MFCD00209607