Order Entry
Italy
ContactUsLinkComponent
Palatinose hydrate ≥98.0% (by HPLC)
Palatinose hydrate ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
Palatinose hydrate ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Melting point:
    118.0 to 127.0 deg-C
  • Specific rotation [a]20/D:
    +97.0 to +101.0 deg (C=1, H2O)
  • Solubility in Water:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together