



Pentylmagnesium bromide 18% ∼1 mol/L in THF
Catalog # TCIAP1177-250G
Supplier: TCI
CAS Number:
Specifications
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Concentration (sec Butanol method):17.0 to 19.8 w/w%
Specification Test Results
About this item
:
ADR:4.3,I
:175.35 g/mol
EINECS:211-747-9
UN:3399
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:693-25-4
Formula:C₅H₁₁BrMg
MDL Number:MFCD00000045