

Propionaldehyde 2,4-dinitrophenylhydrazone ≥97.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:725-00-8
:
Formula:C₉H₁₀N₄O₄
MDL Number:MFCD00191484
ADR:4.1,III
:238.20 g/mol
UN:1325
:GHS02
:GHS07
Specification Test Results
- Appearance:Orange to Brown powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Melting point:148.0 to 155.0 deg-C
Specification Test Results
Frequently Bought Together